SAB5300445

Sigma-Aldrich

Monoclonal Anti-IL10 antibody produced in mouse

clone 3C12C12, ascites fluid

Synonym(s):
IL10
NACRES:
NA.41

Quality Level

biological source

mouse

antibody form

ascites fluid

antibody product type

primary antibodies

clone

3C12C12, monoclonal

species reactivity

human

application(s)

direct ELISA: 1:10,000
western blot: 1:500-1:2,000

isotype

IgG1

conjugate

unconjugated

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... IL-10(3586)

Immunogen

Purified recombinant fragment of human IL-10 expressed in E.coli.
Mouse monoclonal antibody raised against IL-10

Physical form

Ascitic fluid containing 0.03% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

RIDADR

NONH for all modes of transport

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis
Certificate of Origin
Marta Lishnevsky et al.
Experimental and molecular pathology, 97(1), 128-136 (2014-06-28)
Platelet Endothelial Cell Adhesion Molecule 1 (PECAM-1) deficient mice in the FVB/n strain exhibit fatal chronic pulmonary fibrotic disease. The illness occurs in the absence of a detectable pro-inflammatory event. PECAM-1 is vital to the stability of vascular permeability, leukocyte...
Xiaofei Sun et al.
Endocrinology, 155(10), 4006-4014 (2014-07-23)
Infection-induced inflammation, frequently associated with increased production of proinflammatory cytokines, is considered a significant contributor to preterm birth. A G protein-coupled cannabinoid receptor 2 (CB2), encoded by Cnr2, is expressed in various immune cells and was shown to modulate immune...
Qinxiang Zheng et al.
Experimental eye research, 125, 1-8 (2014-05-20)
Tear film hyperosmolarity along with exposure to oxidant stress are factors that can induce chronic ocular surface inflammation and pain. However, there is limited information on how increases in reactive oxygen species (ROS) generated by oxidant exposure can induce inflammation....
Elvira Kurmaeva et al.
Journal of leukocyte biology, 96(3), 377-389 (2014-04-04)
Chronic colitis is accompanied by extensive myelopoiesis and accumulation of CD11b+Gr-1+ cells in spleens and secondary lymphoid tissues. Although cells with similar phenotype have been described in cancer, chronic infection, or autoimmunity, where they were associated with suppression of T...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.