MilliporeSigma
  • SAB5700676
All Photos(5)

SAB5700676

Sigma-Aldrich

Anti-BCL2 antibody produced in rabbit

Synonym(s):
BCL2, Bcl-2, PPP1R50

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

22
26

species reactivity

human, mouse, rat

concentration

3.35 mg/ml

technique(s)

immunofluorescence: 1:50-1:200
immunohistochemistry: 1:50-1:200
immunoprecipitation (IP): 1:50-1:100
western blot: 1:500-1:2000

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... BCL2(596)

Compare Similar Items

View Full Comparison

1 of 4

This Item
HPA055295SAB5701336SAB4500003
biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

-

antibody form

affinity isolated antibody

clone

polyclonal

clone

polyclonal

clone

monoclonal

clone

polyclonal

form

buffered aqueous solution

form

buffered aqueous glycerol solution

form

liquid

form

buffered aqueous solution

mol wt

22, 26

mol wt

-

mol wt

-

mol wt

antigen 26 kDa

species reactivity

human, mouse, rat

species reactivity

human

species reactivity

mouse, rat, human

species reactivity

rat, mouse, human

General description

This gene encodes an integral outer mitochondrial membrane protein that blocks the apoptotic death of some cells such as lymphocytes. Constitutive expression of BCL2, such as in the case of translocation of BCL2 to Ig heavy chain locus, is thought to be the cause of follicular lymphoma. Alternative splicing results in multiple transcript variants.

Immunogen

A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Bcl-2 (NP_000624.2).

Physical form

PBS with 0.02% sodium azide,50% glycerol,pH7.3.

Storage and Stability

Store at -20℃. Avoid freeze / thaw cycles.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service