MilliporeSigma
All Photos(4)

Documents

SAB5701516

Sigma-Aldrich

Anti-THOC4/ALYREF antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

Change View

Select a Size

Change View

biological source

rabbit

Quality Level

antibody product type

primary antibodies

clone

monoclonal

form

liquid

species reactivity

rat, human, mouse

concentration

0.41 mg/mL

technique(s)

immunohistochemistry: 1:50-1:200
western blot: 1:500-1:2000

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ALYREF(10189)

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
SAB5701609SAB5700217SAB5701524
clone

monoclonal

clone

monoclonal

clone

polyclonal

clone

monoclonal

form

liquid

form

liquid

form

buffered aqueous solution

form

liquid

species reactivity

rat, human, mouse

species reactivity

human, rat, mouse

species reactivity

mouse, human, rat

species reactivity

rat, mouse, human

concentration

0.41 mg/mL

concentration

0.43 mg/mL

concentration

0.58 mg/ml

concentration

0.3 mg/mL

technique(s)

immunohistochemistry: 1:50-1:200, western blot: 1:500-1:2000

technique(s)

immunohistochemistry: 1:50-1:200, western blot: 1:500-1:2000

technique(s)

immunofluorescence: 1:50-1:200, western blot: 1:500-1:2000

technique(s)

immunohistochemistry: 1:50-1:200, western blot: 1:500-1:2000

Still not finding the right product?  

Give our Product Selector Tool a try.

General description

The protein encoded by this gene is a heat stable, nuclear protein and functions as a molecular chaperone. It is thought to regulate dimerization, DNA binding, and transcriptional activity of basic region-leucine zipper (bZIP) proteins. [provided by RefSeq, Jul 2008]

Immunogen

A synthesized peptide derived from human THOC4/THOC4/ALYREFREF

Physical form

Buffer: PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service