SAB5702367
Anti-NBS1/NBN Antibody, clone 4G4R1, Rabbit Monoclonal
Synonym(s):
AT-V1, AT-V2, ATV, NBS, NBS1, P95
Sign Into View Organizational & Contract Pricing
Select a Size
Pricing and availability is not currently available.
About This Item
UNSPSC Code:
12352203
NACRES:
NA.46
Skip To
biological source
rabbit
Quality Level
conjugate
unconjugated
material
colorless
clone
4G4R1, monoclonal
form
liquid
mol wt
95 kDa
species reactivity
human
concentration
1.10 mg/mL
technique(s)
immunoprecipitation (IP): 1:50 - 1:200
western blot: 1:500 - 1:2000
color
colorless
isotype
IgG
immunogen sequence
LLTEFRSLVIKNSTSRNPSGINDDYGQLKNFKKFKKVTYPGAGKLPHIIGGSDLIAHHARKNTELEEWLRQEMEVQNQHAKEESLADDLFRYNPYLKRRR
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... NBN(4683)
General description
Mutations in this gene are associated with Nijmegen breakage syndrome, an autosomal recessive chromosomal instability syndrome characterized by microcephaly, growth retardation, immunodeficiency, and cancer predisposition. The encoded protein is a member of the MRE11/RAD50 double-strand break repair complex which consists of 5 proteins. This gene product is thought to be involved in DNA double-strand break repair and DNA damage-induced checkpoint activation. [provided by RefSeq, Jul 2008]
Immunogen
A synthetic peptide corresponding to a sequence within amino acids 655-754 of human NBS1/NBN (O60934).
PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.
Application
WB, IP
Storage Class
10 - Combustible liquids
wgk_germany
WGK 1
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service