Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NA.46
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
2L7W0, monoclonal
Application:
WB
Citations:
-
biological source
rabbit
Quality Segment
conjugate
unconjugated
material
colorless
clone
2L7W0, monoclonal
form
liquid
mol wt
100 kDa
species reactivity
human
concentration
1.50 mg/mL
technique(s)
western blot: 1:500 - 1:2000
color
colorless
isotype
IgG
immunogen sequence
ESDWTENSFQGGQEDGGDDKENHGECKRPWPQHQCSAPADLGYDGRVDSFTDRTSLSPGSQINVLEDPGYLDLTQIEASKPHSAQFLLPVEMFGPGAPKKA
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... PPM1E(22843)
General description
This gene encodes a member of the PPM family of serine/threonine-protein phosphatases. The encoded protein is localized to the nucleus and dephosphorylates and inactivates multiple substrates including serine/threonine-protein kinase PAK 1, 5'-AMP-activated protein kinase (AMPK) and the multifunctional calcium/calmodulin-dependent protein kinases. Alternatively spliced transcript variants have been observed for this gene. [provided by RefSeq, May 2012]
Immunogen
A synthetic peptide corresponding to a sequence within amino acids 500-600 of human PPM1E (Q8WY54).
PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.
Application
WB
Storage Class
10 - Combustible liquids
WGK
WGK 1
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.