Skip to Content
MilliporeSigma

SAB6012209

Anti Ubiquitin Specific Peptidase 28 Antibody, clone 24GB2515, Rabbit Monoclonal

greener alternative

clone 24GB2515, recombinant rabbit monoclonal, expressed in HEK293 cells

Synonym(s):

KIAA1515

Sign In to View Organizational & Contract Pricing.

Select a Size

Pricing and availability is not currently available.

About This Item

UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
24GB2515, recombinant monoclonal
Application:
flow cytometry
immunofluorescence
western blot
Species reactivity:
human, rat
Citations:
Technique(s):
flow cytometry: 1:2000
immunofluorescence: 1:100-:1000
western blot: 1:1,000-1:5,000
Uniprot accession no.:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

Anti Ubiquitin Specific Peptidase 28 Antibody, clone 24GB2515, Rabbit Monoclonal, clone 24GB2515, recombinant rabbit monoclonal, expressed in HEK293 cells

biological source

rabbit (Monoclonal)

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

24GB2515, recombinant monoclonal

form

buffered aqueous glycerol solution

mol wt

122 kDa

species reactivity

human, rat

packaging

vial of 1.5 mL

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

Quality Level

Gene Information

human ... USP28(57646)

sustainability

Greener Alternative Product

concentration

2.005 mg/ml

technique(s)

flow cytometry: 1:2000
immunofluorescence: 1:100-:1000
western blot: 1:1,000-1:5,000

isotype

IgG

Ensembl | human accession no.

NCBI accession no.

UniProt accession no.

greener alternative category

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

The protein encoded by this gene is a deubiquitinase involved in the DNA damage pathway and DNA damage-induced apoptosis. Overexpression of this gene is seen in several cancers. [provided by RefSeq, Oct 2016]
We are committed to bringing you Greener Alternative Products. This product is a Design for Sustainability (DfS) developed product, which belongs to one of the four categories of greener alternatives. Click here to view its DfS scorecard.

Immunogen

A synthesized peptide corresponding to a sequence within amino acids 1020-1077 of human USP28 [NP_065937.1].

Other Notes

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user.

Physical form

Solution in PBS (pH 7.4) containing 50% glycerol, and 0.02% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service