Skip to Content
MilliporeSigma
Get 22% off for Pi Day through March 31.Save Now

SAB6012317

KD-Validated Anti Phospholipase A2 activating protein Antibody, clone 23GB2560, Rabbit Monoclonal

greener alternative

clone 23GB2560, recombinant rabbit monoclonal, expressed in HEK293 cells

Synonym(s):

DOA1, PLA2P, PLAP

Sign In to View Organizational & Contract Pricing.

Select a Size

Pricing and availability is not currently available.

About This Item

UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
23GB2560, recombinant monoclonal
Application:
FACS, IF, WB
Citations:
-

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

biological source

rabbit (Monoclonal)

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

23GB2560, recombinant monoclonal

form

buffered aqueous glycerol solution

mol wt

87 kDa

species reactivity

human

packaging

vial of 1.5 mL

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

concentration

2.275 mg/ml

technique(s)

flow cytometry: 1:2000, immunofluorescence: 1:100-:1000, western blot: 1:1,000-1:5,000

isotype

IgG

Ensembl | human accession no.

NCBI accession no.

UniProt accession no.

greener alternative category

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PLAA(9373)

General description

Predicted to enable ubiquitin binding activity. Involved in cellular response to lipopolysaccharide; macroautophagy; and positive regulation of phospholipase A2 activity. Located in cytoplasm; extracellular exosome; and nucleus. [provided by Alliance of Genome Resources, Dec 2024]
We are committed to bringing you Greener Alternative Products. This product is a Design for Sustainability (DfS) developed product, which belongs to one of the four categories of greener alternatives. Click here to view its DfS scorecard.

Immunogen

A synthesized peptide corresponding to a sequence within amino acids 750-795 of human PLAP [NP_001026859.1].

Physical form

Solution in PBS (pH 7.4) containing 50% glycerol, and 0.02% sodium azide.

Other Notes

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

572.0 °F

flash_point_c

300 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Global Trade Item Number

SKUGTIN
SAB6012317-100UL04065273194852

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service