All Photos(1)

SBP3500058

Sigma-Aldrich

Seasonal H1N1 Hemagglutinin Blocking Peptide for SAB3500058

NACRES:
NA.41

Quality Level

biological source

synthetic

form

liquid

species reactivity

virus

concentration

200 μg/mL

shipped in

wet ice

storage temp.

−20°C

Application

Used as a blocking peptide in immunoblotting applications.

Linkage

This peptide blocks the action of antibody SAB3500058.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service