MilliporeSigma
All Photos(1)

Documents

SCP0185

Sigma-Aldrich

Metalloendopeptidase Substrate

Sign Into View Organizational & Contract Pricing

Empirical Formula (Hill Notation):
C28H37N7O7
Molecular Weight:
583.64
NACRES:
NA.32

assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥75%

storage condition

protect from light

storage temp.

−20°C

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
SCP0101SCP0107SCP0158
Sigma-Aldrich

SCP0185

Metalloendopeptidase Substrate

Sigma-Aldrich

SCP0101

Caspase Substrate

Sigma-Aldrich

SCP0107

Caspase1 Substrate

Sigma-Aldrich

SCP0158

Granzyme B Substrate

form

lyophilized

form

lyophilized

form

lyophilized

form

lyophilized

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage condition

protect from light

storage condition

protect from light

storage condition

protect from light

storage condition

protect from light

composition

Peptide Content, ≥75%

composition

Peptide Content, ≥80%

composition

Peptide Content, ≥85%

composition

Peptide Content, ≥85%

Amino Acid Sequence

Abz-Ala-Gly-Leu-Ala-p-nitro-benzylamide

Application

ABZ-Ala-Gly-Leu-Ala-NBA (AAGLAN) is used to detect metalloendoprotease activity.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Efrat Kessler et al.
Methods in molecular biology (Clifton, N.J.), 1149, 135-169 (2014-05-14)
Pseudomonas aeruginosa secretes into its environment at least seven extracellular proteases: pseudolysin (LasB protease; elastase), aeruginolysin (alkaline proteinase), staphylolysin (staphylolytic endopeptidase; LasA protease), lysyl endopeptidase (protease IV; PrpL), PASP (P. aeruginosa small protease), LepA (Large ExoProtease A), and an aminopeptidase.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service