SML0013
Epinecidine-1 trifluoroacetate salt
≥98% (HPLC)
Synonym(s):
Epinecidin-1 trifluoroacetate salt
About This Item
Skip To
Quality Level
assay
≥98% (HPLC)
form
powder
solubility
H2O: >2 mg/mL
shipped in
wet ice
storage temp.
−20°C
SMILES string
OC(=O)C(F)(F)F.CC[C@H](C)[C@H](NC(=O)[C@H](CCSC)NC(=O)[C@H](CCCCN)NC(=O)CNC(=O)[C@H](C)NC(=O)[C@H](Cc1c[nH]cn1)NC(=O)[C@H](Cc2ccccc2)NC(=O)[C@H](CC(C)C)NC(=O)CNC(=O)[C@H](CCCCN)NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@H](Cc3c[nH]cn3)NC(=O)[C@H](Cc4ccccc4)NC(=O)[C@@H](NC(=O)[C@H](Cc5ccccc5)NC(=O)CN)[C@@H](C)CC)[C@@H](C)CC)[C@@H](C)CC)C(=O)N[C@@H](Cc6c[nH]cn6)C(=O)NCC(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](C(C)C)C(N)=O
InChI
1S/C114H176N30O21S.C2HF3O2/c1-17-67(11)94(113(164)139-86(51-75-55-119-61-125-75)101(152)124-60-92(148)132-82(47-65(7)8)108(159)140-93(66(9)10)98(118)149)141-103(154)80(42-45-166-16)133-102(153)79(41-31-33-44-116)129-90(146)58-122-99(150)71(15)128-104(155)87(52-76-56-120-62-126-76)136-106(157)84(49-73-36-26-22-27-37-73)135-105(156)81(46-64(5)6)131-91(147)59-123-100(151)78(40-30-32-43-115)134-111(162)96(69(13)19-3)144-114(165)97(70(14)20-4)143-110(161)88(53-77-57-121-63-127-77)137-107(158)85(50-74-38-28-23-29-39-74)138-112(163)95(68(12)18-2)142-109(160)83(130-89(145)54-117)48-72-34-24-21-25-35-72;3-2(4,5)1(6)7/h21-29,34-39,55-57,61-71,78-88,93-97H,17-20,30-33,40-54,58-60,115-117H2,1-16H3,(H2,118,149)(H,119,125)(H,120,126)(H,121,127)(H,122,150)(H,123,151)(H,124,152)(H,128,155)(H,129,146)(H,130,145)(H,131,147)(H,132,148)(H,133,153)(H,134,162)(H,135,156)(H,136,157)(H,137,158)(H,138,163)(H,139,164)(H,140,159)(H,141,154)(H,142,160)(H,143,161)(H,144,165);(H,6,7)/t67-,68-,69-,70-,71-,78-,79-,80-,81-,82-,83-,84-,85-,86-,87-,88-,93-,94-,95-,96-,97-;/m0./s1
InChI key
XLUXDFYRYMKELD-GFEQIIKPSA-N
1 of 4
This Item | W3020 | T7830 | D0447 |
|---|---|---|---|
| assay ≥98% (HPLC) | assay ≥98% (HPLC) | assay >98% (HPLC) | assay ≥98% (HPLC) |
| form powder | form powder | form lyophilized powder | form powder |
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. −20°C |
| solubility H2O: >2 mg/mL | solubility H2O: >2 mg/mL | solubility H2O: >2 mg/mL | solubility H2O: ≥2 mg/mL |
| shipped in wet ice | shipped in - | shipped in - | shipped in - |
Application
Biochem/physiol Actions
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service


