SML2697
≥98% (HPLC)
powder
protect from light
white to beige
DMSO: 2 mg/mL, clear
2-8°C
1 of 4
This Item | SML1627 | SML2496 | D6317 |
---|---|---|---|
assay ≥98% (HPLC) | assay ≥98% (HPLC) | assay ≥98% (HPLC) | assay ≥98% (HPLC) |
form powder | form powder | form powder | form solid |
storage condition protect from light | storage condition - | storage condition - | storage condition - |
color white to beige | color yellow to orange | color white to beige | color yellow |
solubility DMSO: 2 mg/mL, clear | solubility DMSO: 20 mg/mL, clear | solubility DMSO: 2 mg/mL, clear | solubility DMSO: ~28 mg/mL, H2O: insoluble |
storage temp. 2-8°C | storage temp. −20°C | storage temp. 2-8°C | storage temp. 2-8°C |
11 - Combustible Solids
WGK 1
Not applicable
Not applicable
Enter Lot Number to search for Certificate of Analysis (COA).
Enter Lot Number to search for Certificate of Origin (COO).
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service