MilliporeSigma
All Photos(2)

Documents

SRP0205

Sigma-Aldrich

MEG-1 Active human

recombinant, expressed in E. coli, ≥90% (SDS-PAGE)

Sign Into View Organizational & Contract Pricing

Synonym(s):
Megakaryocyte phosphatase, PTPMEG1, PTPN4, Protein tyrosine phosphatase, non-receptor type 4
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

assay

≥90% (SDS-PAGE)

form

aqueous solution

mol wt

60.3 kDa

packaging

pkg of 20 μg

concentration

>0.02 mg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PTPN4(5775)

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
SRP0222SRP0212SRP0213
vibrant-m

SRP0205

MEG-1 Active human

vibrant-m

SRP0222

PTPmu Active human

vibrant-m

SRP0212

PTP1B (1-321)/PTPN1 Active human

concentration

>0.02 mg/mL

concentration

>0.02 mg/mL

concentration

0.84 mg/mL

concentration

>0.02 mg/mL

biological source

human

biological source

human

biological source

human

biological source

mouse

assay

≥90% (SDS-PAGE)

assay

≥90% (SDS-PAGE)

assay

≥90% (SDS-PAGE)

assay

≥90% (SDS-PAGE)

NCBI accession no.

NM_002830

NCBI accession no.

BC051651

NCBI accession no.

M31724

NCBI accession no.

NM_011201.3

storage temp.

−70°C

storage temp.

−70°C

storage temp.

−70°C

storage temp.

−70°C

General description

Protein tyrosine phosphatase, Catalytic domain, (amino acds 637-926) (Gene bank accession number NM_002830) with N-terminal GST tag, MW = 60.3 kDa, expressed in an E. coli expression system.

application

Useful for the study of enzyme kinetics, regulation, to dephosphorylate target substrates and for screening inhibitors.

Unit Definition

One unit will hydrolyze 1 nmol p-nitrophenyl phosphate per minute at pH 7.4 and 30°C.

Physical form

Formulated in 25 mM Tris-HCl, pH 8.0, 75 mM NaCl, 0.05% Tween-20, 50% glycerol, 2 mM EDTA, 1 mM DTT, and 10 mM glutathione.

Preparation Note

Thaw on ice. Upon first thaw, briefly spin tube containing enzyme to recover full content of the tube. Aliquot enzyme into single use aliquots. Store remaining undiluted enzyme in aliquots at -70°C. Note: Enzyme is very sensitive to freeze/thaw cycles.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service