MilliporeSigma
All Photos(1)

Documents

SRP2041

Sigma-Aldrich

LXR, β human

recombinant, expressed in E. coli, ≥80% (SDS-PAGE)

Sign Into View Organizational & Contract Pricing

Synonym(s):
LXR-b, LXRB, NER, NER-I, RIP15, UNR
NACRES:
NA.26

biological source

human

recombinant

expressed in E. coli

assay

≥80% (SDS-PAGE)

form

frozen liquid

mol wt

~53 kDa

packaging

pkg of 10 μg

concentration

400 μg/mL

color

clear colorless

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... NR1H2(7376)

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
SRP2130SRP2138SRP2040
vibrant-m

SRP2041

LXR, β human

vibrant-m

SRP2130

LXR, β isoform, GST tagged human

vibrant-m

SRP2040

LXR, α human

recombinant

expressed in E. coli

recombinant

expressed in E. coli

recombinant

expressed in E. coli

recombinant

expressed in E. coli

assay

≥80% (SDS-PAGE)

assay

≥70% (SDS-PAGE)

assay

≥70% (SDS-PAGE)

assay

≥70% (SDS-PAGE)

Gene Information

human ... NR1H2(7376)

Gene Information

human ... NR1H2(7376)

Gene Information

human ... NR1H2(7376)

Gene Information

human ... NR1H3(10062)

shipped in

dry ice

shipped in

dry ice

shipped in

dry ice

shipped in

dry ice

mol wt

~53 kDa

mol wt

~77.5 kDa

mol wt

~55.8 kDa

mol wt

~53.7 kDa

Biochem/physiol Actions

Liver X receptors (LXRs) are nuclear receptors that regulate the metabolism of cholesterol and bile acids. There are two subtypes of LXRs, LXR α and LXR β. LXR α is preferentially expressed in liver, small intestine, kidney and spleen. In contrast, LXR β expression is ubiquitous. The genomic structure and the promoter regions of the two LXR genes contain specific regulatory sites, which suggest that LXRs may have physiological roles in the immune system. Like other nuclear receptors, LXRs heterodimerize with retinoid X receptor (RXR) for function. LXRs are activated by naturally occurring oxysterols and regulate the expression of target genes, including ATP binding cassette transporter 1 (ABC1), ATP binding cassette transporter 8 (ABC8) and cholesterol ester transfer protein (CETP). LXR β expressed in livers of LXR α knockout mice does not compensate for the loss of LXR α. In addition, LXR β, but not LXR α, is also able to activate transcription of a reporter gene, which contains a specific direct repeat separated by 1 bp (DR1) element in the promoter, suggesting that LXR β may have different biological functions.

Physical form

Clear and colorless frozen liquid solution

Preparation Note

Use a manual defrost freezer and avoid repeated freeze-thaw cycles. While working, please keep sample on ice.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

D J Peet et al.
Current opinion in genetics & development, 8(5), 571-575 (1998-10-31)
The liver X receptors (LXRs) are a family of transcription factors that were first identified as orphan members of the nuclear receptor superfamily. The identification of a specific class of oxidized derivatives of cholesterol as ligands for the LXRs has
R Apfel et al.
Molecular and cellular biology, 14(10), 7025-7035 (1994-10-01)
The steroid/hormone nuclear receptor superfamily comprises several subfamilies of receptors that interact with overlapping DNA sequences and/or related ligands. The thyroid/retinoid hormone receptor subfamily has recently attracted much interest because of the complex network of its receptor interactions. The retinoid
P J Willy et al.
Genes & development, 9(9), 1033-1045 (1995-05-01)
We have identified a new retinoid response pathway through which 9-cis retinoic acid (9cRA) activates transcription in the presence of LXR alpha, a member of the nuclear receptor superfamily. LXR alpha shows a specific pattern of expression in visceral organs

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service