MilliporeSigma
All Photos(1)

Documents

T3769

Sigma-Aldrich

Triethylene glycol monooctyl ether

Sign Into View Organizational & Contract Pricing

Synonym(s):
C8E3, Octyl triethylene glycol ether, Octyltriglycol
Linear Formula:
CH3(CH2)7(OCH2CH2)3OH
CAS Number:
Molecular Weight:
262.39
Beilstein/REAXYS Number:
1765631
MDL number:
PubChem Substance ID:

description

non-ionic

mol wt

262.39 g/mol

technique(s)

protein purification: suitable

density

0.948 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

CCCCCCCCOCCOCCOCCO

InChI

1S/C14H30O4/c1-2-3-4-5-6-7-9-16-11-13-18-14-12-17-10-8-15/h15H,2-14H2,1H3

InChI key

XIVLVYLYOMHUGB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Useful in solubilizing membrane proteins.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

J R Elliott et al.
Biochimica et biophysica acta, 814(2), 401-404 (1985-04-11)
The effects of n-decanol, n-hexadecanol, n-octyl(oxyethylene)3 alcohol and cholesterol on gramicidin single-channel lifetime in planar lipid bilayers have been determined. The bilayers used were formed from a solution of monoolein in squalene. Measurements have also been made of the above
Hayter, J.B. and Zulauf, M.
Coll. Polymer Sci., 260, 1023-1023 (1982)
LeMaire, M., et al.
European Journal of Biochemistry, 129, 529-529 (1983)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service