MilliporeSigma
All Photos(1)

T4694

Sigma-Aldrich

Tempidine

≥98% (TLC)

Synonym(s):
2,2,6,6-tetramethylpiperidine hydrochloride, TMP
Empirical Formula (Hill Notation):
C9H19N · HCl
CAS Number:
Molecular Weight:
177.71
MDL number:

assay

≥98% (TLC)

mp

299-301 °C (lit.)

solubility

ethanol: soluble 50 mg/mL
H2O: soluble

SMILES string

Cl.CC1(C)CCCC(C)(C)N1

General description

Antioxidant.

Physical form

Colorless crystal powder

Preparation Note

Tempidine is soluble in water. It dissolves in ethanol at 50 mg/ml.

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service