T9818

Sigma-Aldrich

Trizma® Pre-set crystals

BioPerformance Certified, pH 9.1, average Mw 123.0

Synonym(s):
Tris crystals
Pricing and availability is not currently available.

grade

BioPerformance Certified

form

crystalline powder

mol wt

average Mw 123.0

application(s)

cell culture | mammalian: suitable

impurities

DNase, RNase, Protease, NICKase, none detected
endotoxin, tested
total aerobic microbial count, tested
≤0.05 A290
≤0.5% water (Karl Fischer)
≤5 ppm Heavy metals (as Pb)

pH

8.79 (37 °C, 0.1 M)(lit.)
9.10 (25 °C, 0.1 M)(lit.)
9.1
9.67 (5 °C, 0.1 M)(lit.)

useful pH range

7.0 - 9.0

solubility

water: 0.667 g/mL, clear, colorless

Featured Industry

Diagnostic Assay Manufacturing

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Dissolve in water to make a 0.1M solution. No pH adjustment is necessary. Guaranteed accuracy: ±0.1 pH units.

Tris has a significant temperature coefficient, so select the mixture to give the desired final pH at the desired temperature.
From 5 °C to 25 °C, the pH decreases an average of 0.03 pH units per °C.
From 25 °C to 37 °C, the pH decreases an average of 0.025 pH units per °C.

If a different concentration is desired, note that there is a concentration effect on the pH of the solution.
The product is cell culture tested. It is also tested for low endotoxin levels and analyzed for the absence of nucleases and proteases.

Packaging

100, 250 g in poly bottle

Other Notes

This product is a pre-mixed combination of Trizma base and Trizma HCl that can be used to prepare a common pH value for Tris buffer.
Certain pH electrodes do not give accurate results with Tris buffers.

Preparation Note

Trizma solutions can be autoclaved.

Legal Information

Trizma is a registered trademark of Sigma-Aldrich Co. LLC

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

Hazard Codes

Xi

Risk Statement

36/37/38

Safety Statement

26

RIDADR

NONH for all modes of transport

WGK Germany

2

Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.