MilliporeSigma
All Photos(1)

Documents

UC429

Sigma-Aldrich

Midazolam hydrochloride

solid

Sign Into View Organizational & Contract Pricing

Synonym(s):
8-Chloro-6-(2-fluorophenyl)-1-methyl-4H-imidazo[1,5a][1,4]benzodiazepine hydrochloride
Empirical Formula (Hill Notation):
C18H13ClFN3 · HCl
CAS Number:
Molecular Weight:
362.23
EC Number:
MDL number:
PubChem Substance ID:

form

solid

drug control

USDEA Schedule IV; Home Office Schedule 3; psychotrope (France); kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada; psicótropo (Spain); Decreto Lei 15/93: Tabela IV (Portugal)

color

white to off-white

solubility

H2O: soluble at least 1 mg/mL

storage temp.

2-8°C

SMILES string

Cc1ncc2CN=C(c3ccccc3F)c4cc(Cl)ccc4-n12

InChI

1S/C18H13ClFN3/c1-11-21-9-13-10-22-18(14-4-2-3-5-16(14)20)15-8-12(19)6-7-17(15)23(11)13/h2-9H,10H2,1H3

InChI key

DDLIGBOFAVUZHB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Midazolam has been used as an anesthetic for surgical experiments in rats. This anesthetic has also been used in rats to study the interactions between midazolam and bupivacaine.

Biochem/physiol Actions

Sedative/hypnotic; ligand for the GABAA receptor benzodiazepine modulatory site; CYP3A4 substrate.

Features and Benefits

This compound was developed by Roche. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Preparation Note

Midazolam hydrochloride is soluble in water to at least 1 mg/ml. Product solutions should be protected from light.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Customers Also Viewed

Slide 1 of 5

1 of 5

U Khanderia et al.
Clinical pharmacy, 6(7), 533-547 (1987-07-01)
The chemistry, pharmacology, pharmacokinetics, clinical efficacy, adverse effects, drug interactions, dosage, and cost and availability of midazolam hydrochloride are reviewed. The anxiolytic, sedative, hypnotic, anticonvulsant, muscle-relaxant, and amnesic properties of midazolam are similar to those of other injectable benzodiazepines. Midazolam
Tomoki Nishiyama et al.
Anesthesia and analgesia, 96(5), 1386-1391 (2003-04-23)
Epidurally administered midazolam can potentiate analgesia by epidural bupivacaine. However, whether this effect is synergistic or additive is not known. In this study, we investigated the spinally-mediated analgesic interaction between midazolam and bupivacaine by using the tail-flick and formalin tests
Eduardo Mekitarian Filho et al.
The Journal of pediatrics, 163(4), 1217-1219 (2013-07-03)
This pilot study introduces the aerosolized route for midazolam as an option for infant and pediatric sedation for computed tomography imaging. This technique produced predictable and effective sedation for quality computed tomography imaging studies with minimal artifact and no significant
Natalie L Trevaskis et al.
The Journal of pharmacology and experimental therapeutics, 316(2), 881-891 (2005-10-27)
The influence of the size and turnover kinetics of the enterocyte-based lymph lipid precursor pool (LLPP) on intestinal lymphatic drug transport has been examined. Mesenteric lymph duct-cannulated rats were infused intraduodenally with low (2-5 mg/h) or high (20 mg/h) lipid-dose
D de Wit et al.
Cancer chemotherapy and pharmacology, 73(1), 87-96 (2013-10-24)
Patients treated with sunitinib show substantial inter-patient variability in drug exposure (~30-40 %), which is largely unexplained. Since sunitinib is metabolized by cytochrome P450(CYP)3A4, variability in the activity of this enzyme may explain a considerable proportion of this inter-patient variability.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service