Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V9763

Sigma-Aldrich

V-Pyrro/NO

Synonym(s):

1-[(Ethenyloxy)-NNO-azoxy]-pyrrolidine, O2-Vinyl-1-(pyrrolidin-1-yl)diazen-1-uim-1,2-diolate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11N3O2
Molecular Weight:
157.17
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

storage temp.

−20°C

SMILES string

[O-]\[N+](=N\OC=C)N1CCCC1

InChI

1S/C6H11N3O2/c1-2-11-7-9(10)8-5-3-4-6-8/h2H,1,3-6H2/b9-7+

InChI key

RJWXCQACZTVGIN-VQHVLOKHSA-N

Biochem/physiol Actions

Liver-selective nitric oxide donor that blocks TNF-α-induced apoptosis and toxicity

pictograms

Flame

signalword

Danger

hcodes

Hazard Classifications

Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

57.2 °F - closed cup

flash_point_c

14 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service