MilliporeSigma
All Photos(6)

Documents

WH0000053M1

Sigma-Aldrich

Monoclonal Anti-ACP2 antibody produced in mouse

clone M1-4A12, purified immunoglobulin, buffered aqueous solution

Sign Into View Organizational & Contract Pricing

Synonym(s):
Anti-acid phosphatase 2, lysosomal
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

M1-4A12, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
indirect ELISA: suitable
indirect immunofluorescence: suitable
western blot: 1-5 μg/mL

isotype

IgG2bλ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ACP2(53)

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
SAB1410966SAB1403518WH0000052M1
conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

biological source

mouse

biological source

rabbit

biological source

mouse

biological source

mouse

Gene Information

human ... ACP2(53)

Gene Information

human ... ACP2(53)

Gene Information

human ... ACP1(52)

Gene Information

human ... ACP1(52)

clone

M1-4A12, monoclonal

clone

polyclonal

clone

2A3, monoclonal

clone

4B10, monoclonal

species reactivity

human

species reactivity

human

species reactivity

mouse, human, rat

species reactivity

mouse, rat, human

Still not finding the right product?  

Give our Product Selector Tool a try.

General description

This gene encodes the beta subunit of lysosomal acid phosphatase (LAP). LAP is chemically and genetically distinct from red cell acid phosphatase. The encoded protein belongs to a family of distinct isoenzymes which hydrolyze orthophosphoric monoesters to alcohol and phosphate. Mutations in this gene or in the related alpha subunit gene cause acid phosphatase deficiency. Multiple alternatively spliced transcript variants encoding different isoforms have been identified for this gene. (provided by RefSeq)

Immunogen

ACP2 (AAH03160, 1 a.a. ~ 423 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MAGKRSGWSRAALLQLLLGVNLVVMPPTQARSLRFVTLLYRHGDRSPVKTYPKDPYQEEEWPQGFGQLTKEGMLQHWELGQALRQRYHGFLNTSYHRQEVYVRSTDFDRTLMSAEANLAGLFPPNGMQRFNPNISWQPIPVHTVPITEDRLLKFPLGPCPRYEQLQNETRQTPEYQNESSRNAQFLDMVANETGLTDLTLETVWNVYDTLFCEQTHGLRLPPWASPQTMQRLSRLKDFSFRFLFGIYQQAEKARLQGGVLLAQIRKNLTLMATTSQLPKLLVYSAHDTTLVALQMALDVYNGEQAPYASCHIFELYQEDSGNFSVEMYFRNESDKAPWPLSLPGCPHRCPLQDFLRLTEPVVPKDWQQECQLASGPADTEVIVALAVCGSILFLLIVLLLTVLFRMQAQPPGYRHVADGEDHA

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service