Select a Size
$580.00
$580.00
About This Item
Skip To
biological source
mouse
Quality Level
conjugate
unconjugated
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
2G10, monoclonal
form
buffered aqueous solution
species reactivity
human
technique(s)
immunofluorescence: suitable
indirect ELISA: suitable
isotype
IgG1κ
GenBank accession no.
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... HDAC9(9734)
1 of 4
This Item | H6287 | AV42788 | WH0055869M1 |
|---|---|---|---|
| antibody form purified immunoglobulin | antibody form purified immunoglobulin | antibody form IgG fraction of antiserum | antibody form purified immunoglobulin |
| species reactivity human | species reactivity mouse, human | species reactivity sheep, human, guinea pig, mouse | species reactivity rat, mouse, human |
| conjugate unconjugated | conjugate unconjugated | conjugate unconjugated | conjugate unconjugated |
| biological source mouse | biological source mouse | biological source rabbit | biological source mouse |
| Gene Information human ... HDAC9(9734) | Gene Information human ... HDAC1(3065) | Gene Information human ... HDAC9(9734) | Gene Information human ... HDAC8(55869) |
| clone 2G10, monoclonal | clone HDAC1-21, monoclonal | clone polyclonal | clone 2F4, monoclonal |
General description
Immunogen
Sequence
QIHMNKLLSKSIEQLKQPGSHLEEAEEELQGDQAMQEDRAPSSGNSTRSDSSACVDDTLGQVGAVKVKEEPVDSDEDAQIQEMESGEQAA
Physical form
Legal Information
Not finding the right product?
Try our Product Selector Tool.
Storage Class
10 - Combustible liquids
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service


