Z330116
low-density polyethylene bottle
round bottle
translucent bottle
wide-mouth bottle
Joints threaded neck (GL50)
with cap
pkg of 50 ea
BRAND 129654
500 mL
76 mm × 155 mm
natural
Looking for similar products? Visit Product Comparison Guide
1 of 4
This Item | Z330132 | Z329959 | Z329967 |
---|---|---|---|
material low-density polyethylene bottle, round bottle, translucent bottle, wide-mouth bottle | material low-density polyethylene bottle, round bottle, translucent bottle, wide-mouth bottle | material low-density polyethylene bottle, narrow-mouth bottle, round bottle, translucent | material low-density polyethylene bottle, narrow-mouth bottle, round bottle, translucent |
joint Joints threaded neck (GL50) | joint Joints threaded neck (GL65) | joint Joints threaded neck (GL18) | joint Joints threaded neck (GL18) |
feature with cap | feature with cap | feature with cap | feature with cap |
packaging pkg of 50 ea | packaging pkg of 25 ea | packaging pkg of 100 | packaging pkg of 50 |
manufacturer/tradename BRAND 129654 | manufacturer/tradename BRAND 129664 | manufacturer/tradename BRAND 129428 | manufacturer/tradename BRAND 129438 |
capacity 500 mL | capacity 2,000 mL | capacity 50 mL | capacity 100 mL |
Enter Lot Number to search for Certificate of Analysis (COA).
Enter Lot Number to search for Certificate of Origin (COO).
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service