MilliporeSigma
Skip To
Z332240
Bottle adapters for BRAND® Dispensers and Burettes
GL 32 thread, GL 45 bottle, polypropylene
Synonym(s):
Bottle adapter for Dispensette/ seripettor / Titrette / QuikSip
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Size/SKUAvailabilityPrice
1 ea
Estimated to ship onMay 26, 2026fromMILWAUKEE
$14.20
About This Item
NACRES:
NB.01
UNSPSC Code:
41121815
$14.20
Estimated to ship onMay 26, 2026Details
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
material
polypropylene
manufacturer/tradename
BRAND 704345
fitting
GL 45 bottle, GL 32 thread
General description
For all Dispensette® models as well as seripettor® and QuikSip
Other Notes
BR704345 (India)
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Dispensette is a registered trademark of BRAND GMBH + CO KG
QuikSip is a trademark of BRAND GMBH + CO KG
seripettor is a registered trademark of BRAND GMBH + CO KG
Compare Similar Items
View Full Comparison
Show Differences
1 of 1
This Item
Z332305Z332283Z332291
material
polypropylene
material
polypropylene
material
polypropylene
material
polypropylene
fitting
GL 45 bottle, GL 32 thread
fitting
GL 38 bottle, GL 45 thread
fitting
GL 32 bottle, GL 45 thread
fitting
S 40 bottle, GL 45 thread
manufacturer/tradename
BRAND 704345
manufacturer/tradename
BRAND 704397
manufacturer/tradename
BRAND 704396
manufacturer/tradename
BRAND 704343
Still not finding the right product?
Choose from one of the most recent versions:
Certificates of Analysis (COA)