Documents
Z338923
Sigma-Aldrich
Comb for mini horizontal electrophoresis unit
size 1.5 mm × 12 wells
NACRES:
NB.22
feature
well spacing compatible with multichannel pipettes
size
1.5 mm × 12 wells
Certificate of Analysis
Certificate of Origin
Contact Technical Service
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.