Documents
Z339466
Sigma-Aldrich
Maxi horizontal electrophoresis unit accessory
Colored gel loading strips
NACRES:
NB.22
Compatibility
Provide a colored background behind sample loading area making sample wells more visible. PAK of 12, 4 colors
Certificate of Analysis
Certificate of Origin
Contact Technical Service
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.