Skip to Content
MilliporeSigma

Z4626

ZM 39923 hydrochloride

>98% (HPLC)

Synonym(s):

3-(N-Benzyl-N-isopropyl)amino-1-(naphthalen-2-yl)propan-1-one hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

Empirical Formula (Hill Notation):
C23H25NO · HCl
CAS Number:
Molecular Weight:
367.91
UNSPSC Code:
12352200
PubChem Substance ID:
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

ZM 39923 hydrochloride, >98% (HPLC)

InChI

1S/C23H25NO.ClH/c1-18(2)24(17-19-8-4-3-5-9-19)15-14-23(25)22-13-12-20-10-6-7-11-21(20)16-22;/h3-13,16,18H,14-15,17H2,1-2H3;1H

SMILES string

Cl[H].CC(C)N(CCC(=O)c1ccc2ccccc2c1)Cc3ccccc3

InChI key

NJTUORMLOPXPBY-UHFFFAOYSA-N

assay

>98% (HPLC)

form

solid

color

white

mp

151-154 °C (lit.)

solubility

DMSO: soluble 16 mg/mL

storage temp.

2-8°C

Gene Information

human ... JAK3(3718)

Biochem/physiol Actions

Selective JAK3 inhibitor.

Packaging

Photosensitive and hygroscopic.

Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service