Z4875

Sigma-Aldrich

Zinc chloride

≥98%

Synonym(s):
Dichlorozinc
Linear Formula:
ZnCl2
CAS Number:
Molecular Weight:
136.30
EC Number:
MDL number:

vapor pressure

1 mmHg ( 428 °C)

assay

≥98%

mp

293 °C (lit.)

SMILES string

[Cl-].[Cl-].[Zn++]

InChI

1S/2ClH.Zn/h2*1H;/q;;+2/p-2

InChI key

JIAARYAFYJHUJI-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

UN 2331 8 / PGIII

WGK Germany

3

L Davies et al.
Cell death & disease, 2, e136-e136 (2011-04-01)
The activation and regulation of target genes by the tumour-suppressor p53 dictates the fate of a cell, with cell cycle arrest or apoptosis being two distinct outcomes. PERP (p53 apoptosis effector related to PMP-22), a p53 transcriptional target, is induced...
İbrahim Gülseren et al.
Food chemistry, 135(2), 770-774 (2012-08-08)
Whey protein isolate (WPI) nanoparticles were prepared using ethanol desolvation, and their capacity to incorporate ZnCl(2) was analysed. Desolvation was carried out at pH 9 and the volume of added ethanol was 0-3 times the volume of protein solution. The...
Manuella P Kaster et al.
Pharmacological reports : PR, 64(3), 706-713 (2012-07-21)
In this work, the contribution of NMDA receptors to the antidepressant-like effect of adenosine in the forced swimming test (FST) was investigated. The pretreatment of mice with NMDA or D-serine was able to prevent the anti-immobility effect of either adenosine...
Kaushik Nath et al.
Environmental science and pollution research international, 20(6), 4030-4045 (2012-12-06)
Utilization of agrowaste materials for the production of activated carbon, as an excellent adsorbent with large surface area, is well established industrially, for dephenolation of wastewater. In the present work, dried pods of Prosopis cineraria-a novel and low-cost agrowaste material-were...
Andrea M Hui et al.
Dermatologic clinics, 30(3), 503-515 (2012-07-18)
For melanoma treatment, the primary goal is complete removal with histologically negative margins. Mohs micrographic surgery (MMS) has been extensively used and studied for the treatment of nonmelanoma skin cancer, particularly at sites where tissue conservation is vital. The use...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.