Documents
Z740249
Dymo® LabelWriter® 450
pkg of 1 ea
packaging
pkg of 1 ea
compatibility
For the Related labels to print correclty they must be purchased from a US supplier.
Related Categories
General description
The Dymo® LabelWriter® 450 printer is a direct thermal printer and does not use ink or toner. Instead, the printer uses heat from a thermal print head to print on specially treated, heat-sensitive labels. Labels are the only supplies you will ever need to buy.
System Requirements.
The LabelWriter® printer requires a USB 1.1 or USB 2.0 connection with one of the following:
  • Windows XP 32-bit
  • Windows 7
To use the LabelWriter® SE450 printer with a serial connection, an RS-232 serial port is required.
Please note: For the ′Related′ labels to print correclty they must be purchased from a US supplier.
Legal Information
Dymo is a registered trademark of Diversified Biotech
LabelWriter is a registered trademark of Diversified Biotech
Certificate of Analysis
Certificate of Origin
Contact Technical Service
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.