Documents
Z748374
3DKUBE Preload Biomerix 3D Scaffold Disks, Biomerix Biomaterial in 3DKUBE independent chambers
Synonym(s):
3D Cell Culture, 3D
Pricing and availability is not currently available.
mfr. no.
Kiyatec, PL021
General description
Biomerix Biomaterial Scaffold Disks:
  • Polycarbonate Polyurethane-urea
  • Interconnected Porosity, 90-95% Void Volume
  • Designed to Mimic Nature & Function of ECM
3DKUBE Plasticware:
  • Independent Chambers Configuration
  • 4 Preloaded Scaffolds per Chamber
  • 4 Assembled 3DKUBEs per Case
Legal Information
3DKUBE is a trademark of Kiyatec, Inc.
Andrew Ward et al.
PloS one, 8(2), e55696-e55696 (2013-02-14)
The efficacy and economy of most in vitro human models used in research is limited by the lack of a physiologically-relevant three-dimensional perfused environment and the inability to noninvasively quantify the structural and biochemical characteristics of the tissue. The goal...
The 3DKUBE Segregated Co-Culture System: Optimal Tumor-Stromal Interactions For Modeling The Tumor Microenvironment In Anti-cancer Therapeutic Development.
Crosswell H, Widener RD, Desgupta A, Shuford S, Gevaert MR, Orr DE, Langenheim J.
Cancer Research, 73, 1538-7445 (2013)
The Roles Of Various Metabolic Enzymes In Diabetic Cardiomyopathy - In Vivo And In Vitro Approach.
Larrew T, Chow J, Schulte J, Simionescu D, Simionescu A.
Cardiovascular Pathology : the Official Journal of the Society For Cardiovascular Pathology, 22(3), e48-e48 (2013)
Karolina Janeczek Portalska et al.
Journal of tissue engineering and regenerative medicine, 10(5), 363-373 (2013-04-18)
Modular tissue engineering is a strategy to create scalable, self-assembling, three-dimensional (3D) tissue constructs. This strategy was used to deliver endothelial-like cells derived from bone marrow mesenchymal stromal cells (EL-MSCs) to locally induce vascularization. First, tissue engineered modules were formed...
Sayed-Hadi Mirmalek-Sani et al.
Biomaterials, 34(22), 5488-5495 (2013-04-16)
Emergent technologies of regenerative medicine have the potential to overcome the limitations of organ transplantation by supplying tissues and organs bioengineered in the laboratory. Pancreas bioengineering requires a scaffold that approximates the biochemical, spatial and vascular relationships of the native...
Contact Technical Service
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.