Documents
Z756814
Equilibrium type dialysis cell
capacity 1 mL, pack of 1 ea
Synonym(s):
dialysis tubes
NACRES:
NB.22
material
clear acrylic
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art H40260-0000
capacity
1 mL
pore size
≥6 kDa MWCO
General description
These dialysis units feature stainless steel screws which secure the membrane acting as a seal. PTFE-coated stainless steel screws are located in the filler holes. Volume displayed is for a single half-cell. Total cell capacity is 2x of that shown. A red dot is featured on one half of the dialysis cell for easily identifying otherwise identical halves.
Certificates of Analysis (COA)
Already Own This Product?
Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.
Visit the Document Library
Difficulty Finding Your Product Or Lot/Batch Number?
Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).
Example:
T1503
Product Number
-
25G
Pack Size/Quantity
Additional examples:
705578-5MG-PW
PL860-CGA/SHF-1EA
MMYOMAG-74K-13
1000309185
enter as 1.000309185)
Having trouble? Feel free to contact Technical Service for assistance.
Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.
Aldrich Products
  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').
  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').
  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.
Not Finding What You Are Looking For?
In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.
Request COA