120 V AC, US 3-pin plug
to hold 4 8 x 0.2mL strip tubes
to hold 8 1.5/2.0 mL microcentrifuge tubes
Labnet C1801
2000 × g
6000 rpm
centrifugation: suitable
Looking for similar products? Visit Product Comparison Guide
1 of 4
This Item | Z681717 | Z763136 | Z763144 |
---|---|---|---|
AC/DC input 120 V AC, US 3-pin plug | AC/DC input - | AC/DC input - | AC/DC input - |
feature to hold 4 8 x 0.2mL strip tubes, to hold 8 1.5/2.0 mL microcentrifuge tubes | feature to hold 12 1.5/2.0 mL tubes, to hold 32 0.2 mL PCR tubes, to hold 4 8-tube PCR strips, 0.2 mL | feature Holds 0.4 mL tubes | feature Holds 0.5 mL tubes |
manufacturer/tradename Labnet C1801 | manufacturer/tradename Benchmark Scientific C1012 | manufacturer/tradename Labnet C1206-8 | manufacturer/tradename Labnet C1205-8 |
parameter 2000 × g, 6000 rpm | parameter 2000 × g, 5500 rpm speed (2000 × g) | parameter - | parameter - |
technique(s) centrifugation: suitable | technique(s) PCR: suitable, centrifugation: suitable | technique(s) centrifugation: suitable | technique(s) centrifugation: suitable |
Enter Lot Number to search for Certificate of Analysis (COA).
Enter Lot Number to search for Certificate of Origin (COO).
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service