ZRB1658
Anti-p-Paxillin-Tyr118 Antibody, clone 1E19 ZooMAb® Rabbit Monoclonal

recombinant, expressed in HEK 293 cells
Skip To
biological source
rabbit
Quality Level
recombinant
expressed in HEK 293 cells
conjugate
unconjugated
antibody form
purified antibody
antibody product type
primary antibodies
clone
1E19, recombinant monoclonal
description
recombinant, expressed in HEK 293 cells
product line
ZooMAb® learn more
form
lyophilized
mol wt
calculated mol wt 64.51 kDa
observed mol wt ~70 kDa
purified by
using Protein A
species reactivity
mouse
species reactivity (predicted by homology)
human, rat
packaging
antibody small pack of 25 μL
greener alternative product characteristics
Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.
enhanced validation
recombinant expression
Learn more about Antibody Enhanced Validation
sustainability
Greener Alternative Product
technique(s)
flow cytometry: suitable
immunocytochemistry: suitable
inhibition assay: suitable
western blot: suitable
isotype
IgG
epitope sequence
N-terminal
Protein ID accession no.
UniProt accession no.
greener alternative category
shipped in
ambient
storage temp.
2-8°C
target post-translational modification
phosphorylation (pTyr118)
Gene Information
human ... PXN(5829)
1 of 4
This Item | ZRB1284 | ZRB1794 | ZRB1736 |
|---|---|---|---|
| antibody form purified antibody | antibody form purified antibody | antibody form purified antibody | antibody form purified antibody |
| species reactivity mouse | species reactivity human, mouse | species reactivity mouse, human, monkey | species reactivity mouse, human |
| conjugate unconjugated | conjugate unconjugated | conjugate unconjugated | conjugate unconjugated |
| Gene Information human ... PXN(5829) | Gene Information - | Gene Information human ... USP7(7874) | Gene Information human ... ZNF683(257101) |
| biological source rabbit | biological source rabbit | biological source rabbit | biological source rabbit |
| clone 1E19, recombinant monoclonal | clone 1L19, recombinant monoclonal | clone 1E19, recombinant monoclonal | clone 1N19, recombinant monoclonal |
General description
Immunogen
Application
Evaluated by Western Blotting in lysates from C2C12 cells treated with Sodium Pervanadate.
Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Paxillin phosphorylated on tyrosine 118 in lysate from overnight serum starved C2C12 cells treated with 5 mM sodium pervanadate, but not in untreated cells.
Tested applications
Flow Cytometry Analysis: 0.1 μg from a representative lot detected p-Paxillin-Tyr118 in untreated and Sodium Pervanadate treated C2C12 cells.
Peptide Inhibition Assay: Target band detection in a lysate from C2C12 cells treated sodium pervanadate was prevented by preblocking of a representative lot with the immunogen phosphopeptide, but not the corresponding non-phosphopeptide.
Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected p-Paxillin-Tyr118 in C2C12 treated with pervanadate cell line.
Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Biochem/physiol Actions
Physical form
Preparation Note
Legal Information
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service


