Skip to Content
MilliporeSigma

ZRB2012

Anti-p70S6 Kinase Antibody, clone 1E13 ZooMAb® Rabbit Monoclonal

greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Ribosomal protein S6 kinase beta-1;EC 2.7.11.1;S6K-beta-1;S6K1;70 kDa ribosomal protein S6 kinase 1;P70S6K1;p70-S6K 1;Ribosomal protein S6 kinase I;Serine/threonine-protein kinase 14A;p70 ribosomal S6 kinase alpha; p70 S6 kinase alpha;p70 S6K-alpha;p70S6K


Sign In to View Organizational & Contract Pricing.

Select a Size

Pricing and availability is not currently available.

About This Item

NACRES:
NA.43
UNSPSC Code:
12352203

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

Anti-p70S6 Kinase Antibody, clone 1E13 ZooMAb® Rabbit Monoclonal, recombinant, expressed in HEK 293 cells

biological source

rabbit

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1E13, recombinant monoclonal

description

1E13 Clone

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 59.14 kDa
observed mol wt ~60 kDa

purified by

using Protein A

species reactivity

human, monkey

species reactivity (predicted by homology)

mouse, rat

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Quality Level

Gene Information

human ... RPS6KB1(6198)

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
ZRB1948ZRB1669ZRB1901
clone

1E13, recombinant monoclonal

clone

1C18, recombinant monoclonal

clone

1I22, recombinant monoclonal

clone

2H10, recombinant monoclonal

Gene Information

human ... RPS6KB1(6198)

Gene Information

human ... RPS6KA1(6195)

Gene Information

human ... RPS6(6194)

Gene Information

human ... RPS6(6194)

antibody form

purified antibody

antibody form

purified antibody

antibody form

purified antibody

antibody form

purified antibody

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

species reactivity

human, monkey

species reactivity

rat, human

species reactivity

human

species reactivity

mouse, human

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

Preparation Note

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.
Reconstitute lyophilized antibody pellet with 25μL of ultrapure water or Phosphate Buffered Saline (PBS). Please refer to our reconstitution protocol and the specific application guidance on the suggested starting dilutions and sample type.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

General description

Ribosomal protein S6 kinase beta-1 (UniProt: P23443; also known as EC 2.7.11.1, S6K-beta-1, S6K1, 70 kDa ribosomal protein S6 kinase 1, P70S6K1, p70-S6K 1, Ribosomal protein S6 kinase I, Serine/threonine-protein kinase 14A, p70 ribosomal S6 kinase alpha, p70 S6 kinase alpha, p70 S6K-alpha, p70 S6KA) is encoded by the RPS6KB1 (also known as STK14A) gene (Gene ID: 6198) in human. p70S6K1 is a serine/threonine-protein kinase that acts downstream of mTOR signaling in response to growth factors and nutrients to promote cell proliferation, cell growth, and cell cycle progression. It regulates protein synthesis through phosphorylation of EIF4B, RPS6, and EEF2K and contributes to cell survival by repressing the pro-apoptotic function of BAD. Upon mitogenic stimulation, phosphorylation by mTORC1 leads to its activation following dissociation from the EIF3 complex. The active p70S6K1 then phosphorylates and activates several substrates in the pre-initiation complex. It is also shown to play a role in feedback regulation of mTORC2 by mTORC1 by phosphorylating RICTOR, which results in the inhibition of mTORC2 and Akt1 signaling. The protein kinase domain of p70S6K1 is localized to amino acids 91-352 and the autoinhibitory domain is localized to amino acids 424-525. The autoinhibitory domain blocks phosphorylation within the AGC-kinase C-terminal domain and the activation loop. Its activation requires multiple phosphorylation events on serine/threonine residues. Phosphorylation of multiple sites in the autoinhibitory domain disrupts the autoinhibitory mechanism and allows phosphorylation of threonine 252 by PDPK1. The active conformation of the kinase is stabilized by phosphorylation of sites (Thr 252; Ser 394; and Thr 412) located in the kinase domain activation loop. It also contains a TOR signaling (TOS) motif (aa 28-32) that is essential for its activation by mTORC1. Five isoforms of p70S6K1 have been described that are produced by alternative splicing and alternative initiation. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Wang, X., et al. (2001). EMBO J. 20(16); 4370-4379; Pullen, N., et al. (1998). Science. 279(5351); 707-710).
We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Immunogen

KLH-conjugated linear peptide corresponding to 21 amino acids from the N-terminal region of human p70S6 Kinase.

Application

Quality Control Testing

Evaluated by Western Blotting in COS-7 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected p70S6 Kinase in COS-7 cell lysate.

Tested Applications

Western Blotting Analysis: A 1:10,000 dilution from a representative lot detected p70S6 Kinase in A431 cell lysate.

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected p70S6 Kinase in human bone marrow tissue sections.

Affinity Binding Assay: A representative lot of this antibody bound p70 S6 Kinase peptide with a KD of 8.9 x 10-6 in an affinity binding assay.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected p70S6 Kinase in A431 cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Biochem/physiol Actions

Clone 1E13 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects p70S6 kinase. It targets an epitope within 21 amino acids from the N-terminal region.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service