Skip to Content
MilliporeSigma

ZRB2392

Anti-MARC1 Antibody, clone 1G13 ZooMAb® Rabbit Monoclonal

greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

MOSC domain-containing protein 1, Moco sulfurase C-terminal domain-containing protein 1, Mitochondrial amidoxime-reducing component 1, Molybdenum cofactor sulfurase C-terminal domain-containing protein 1, mARC1


Sign In to View Organizational & Contract Pricing.

Select a Size

Pricing and availability is not currently available.

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

Anti-MARC1 Antibody, clone 1G13 ZooMAb® Rabbit Monoclonal, recombinant, expressed in HEK 293 cells

biological source

rabbit

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1G13, recombinant monoclonal

description

1G13 Clone

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 37.5 kDa
observed mol wt ~38 kDa

purified by

using Protein A

species reactivity

rat, mouse, human

species reactivity (predicted by homology)

porcine, monkey

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

Internal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Quality Level

Gene Information

human ... MTARC1(64757)

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
ZRB1976ZRB2365ZRB2328
conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

clone

1G13, recombinant monoclonal

clone

1B13, recombinant monoclonal

clone

1F4, recombinant monoclonal

clone

1K19, recombinant monoclonal

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

species reactivity

rat, mouse, human

species reactivity

human

species reactivity

human

species reactivity

human, mouse, rat

Gene Information

human ... MTARC1(64757)

Gene Information

human ... RIGI(23586)

Gene Information

human ... ZC3H13(23091)

Gene Information

human ... MBD3(53615)

antibody form

purified antibody

antibody form

purified antibody

antibody form

purified antibody

antibody form

purified antibody

Preparation Note

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.
Reconstitute lyophilized antibody pellet with 25μL of ultrapure water or Phosphate Buffered Saline (PBS). Please refer to our reconstitution protocol and the specific application guidance on the suggested starting dilutions and sample type.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

General description

ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.
Mitochondrial amidoxime-reducing component 1 (UniProt: Q5VT66; also known as mARC1, Molybdenum cofactor sulfurase C-terminal domain-containing protein 1, MOSC domain-containing protein 1, Moco sulfurase C-terminal domain-containing protein 1) is encoded by the MTARC1 (also known as MARC1, MOSC1) gene (Gene ID: 64757) in human. MARC1 is a single-pass type II outer mitochondrial membrane protein that consists of a mitochondrial matrix domain (aa 2-20), a transmembrane domain (aa 21-40), and a cytoplasmic domain (aa 41-337). It catalyzes the reduction of N-oxygenated molecules, acting as a counterpart of cytochrome P450 and flavin-containing monooxygenases in metabolic cycles. It is known to reduce a multitude of N-hydroxylated prodrugs, particularly amidoximes, leading to increased drug bioavailability. It is also reported to be involved in mitochondrial N(omega)-hydroxy-L-arginine (NOHA) reduction, thereby regulating endogenous nitric oxide levels and biosynthesis. It has two structural domains, the molybdenum cofactor(Moco) sulfurase C-terminal (MOSC) domain (aa 187-335) and the MOSC N-terminal region (aa 93-183), forming a cleft that accommodates Moco. The MOSC domain that contains a large seven-stranded mostly antiparallel beta-barrel, engages multiple interactions with Moco pterin ring and phosphate group, which allows for a tight coordination of Moco within the core of the enzyme. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Kubitza, C., et al. (2018). Proc. Natl. Acad. Sci. USA. 115(47); 11958-11963; Krompholz, N., et al. (2012). Chem. Res. Toxicol. 25(11); 2443-2450; Kotthaus, J., et al. (2011). Biochem. J. 433(2); 383-391).
We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry. This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.

Immunogen

KLH-conjugated linear peptide corresponding to 16 amino acids from the internal region, cytoplasmic domain of human Mitochondrial amidoxime-reducing component 1 (MARC1).

Application

Quality Control Testing

Evaluated by Western Blotting in Human liver tissue lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected MARC1 in Human liver tissue lysate.

Tested Applications

Western Blotting Analysis: A 1:10,000 dilution from a representative lot detected MARC1 in Mouse liver and Rat liver tissue lysates.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected MARC1 in HepG2 cells.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected MARC1 in human liver tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Biochem/physiol Actions

Clone 1G13 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects MARC1. It targets an epitope within 16 amino acids from the internal region.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service