Skip to Content
MilliporeSigma
All Photos(6)

Key Documents

ZRB2880

Anti-GRK2 Antibody, clone 2C6 ZooMAb® Rabbit Monoclonal

newenhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

ADRBK1, BARK1, Beta-ARK-1, Beta-adrenergic receptor kinase 1, EC:2.7.11.15, G-protein coupled receptor kinase 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

2C6, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 79.57 kDa
observed mol wt ~80 kDa

purified by

using Protein A

species reactivity

human, rat, mouse

packaging

antibody small pack of 25

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal half

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... GRK2(156)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of the 12 Principles of Green Chemistry. This antibody is preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed, and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 2C6 is a ZooMAb® rabbit recombinant monoclonal antibody that detects GRK2. It targets an epitope within 23 amino acids from the N-terminal half.

Immunogen

KLH-conjugated linear peptide corresponding to 23 amino acids from the N-terminal half of human GRK2.

Application

Quality Control Testing

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: A 1:10,000 dilution of this antibody detected GRK2 in HeLa cell lysate.

Tested Applications

Western Blotting Analysis: A 1:1,000-10,000 dilution from a representative lot detected GRK2 in lysates from THP-1 cells, mouse brain tissue, and rat spleen tissue, human spleen tissue lysate.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected GRK2 in human spleen tissue sections.

Affinity Binding Assay: A representative lot of this antibody bound with GRK2 peptide with at least one hundred thousand-fold (100,000X) higher affinity than with non-specific control peptide.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Beta-adrenergic receptor kinase 1 (UniProt: P25098; EC:2.7.11.15 also known as Beta-ARK-1, G-protein coupled receptor kinase 2) is encoded by the GRK2 (also known as ADRBK1, BARK, BARK1) gene (Gene ID: 156) in human. GRK2 is a member of the G protein-coupled receptor kinase (GRK) family. It consists of an N-terminal domain that interacts with G protein-coupled receptors (GPCRs), a central catalytic domain responsible for its kinase activity, and a C-terminal domain that mediates interactions with other proteins and lipids. GRK2 plays a crucial role in the regulation of GPCR signaling. Upon activation of GPCRs, GRK2 phosphorylates specific serine and threonine residues on the receptor, leading to the recruitment of arrestin proteins. This process desensitizes the receptor, preventing further G protein activation and initiating receptor internalization and downregulation. GRK2 thus modulates the intensity and duration of GPCR signaling, impacting various physiological processes. GRK2 is implicated in several diseases. In cardiovascular diseases, such as heart failure, elevated levels of GRK2 are associated with impaired β-adrenergic receptor signaling, contributing to decreased cardiac function. Additionally, GRK2 has been implicated in metabolic disorders, inflammatory diseases, and certain cancers, where its dysregulation can alter cellular signaling pathways, affecting disease progression and response to treatment. Modulating GRK2 activity is being explored as a therapeutic strategy in these conditions. This ZooMAb® recombinant monoclonal antibody, generated by our proprietary technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service