Skip to Content
MilliporeSigma

ZRB3462

Anti-OLR1 Antibody, clone 2E2 ZooMAb® Rabbit Monoclonal

recombinant, expressed in HEK 293 cells

Synonym(s):

C-type lectin domain family 8 member A, CLEC8A, LOX-1, Lectin-like oxLDL receptor 1, Lectin-like oxidized LDL receptor 1, Lectin-type oxidized LDL receptor 1, Ox-LDL receptor 1, Oxidized low-density lipoprotein receptor 1, Oxidized low-density lipoprotein receptor 1, soluble form, hLOX-1

Sign In to View Organizational & Contract Pricing.

Select a Size

Pricing and availability is not currently available.

About This Item

UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
2E2, recombinant monoclonal
Application:
IHC, WB, affinity binding assay
Species reactivity:
human, mouse, rat
Citations:
Technique(s):
affinity binding assay: suitable, immunohistochemistry: suitable, western blot: suitable
Uniprot accession no.:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

biological source

rabbit

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

2E2, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 30.96 kDa, observed mol wt ~52 kDa

purified by

using Protein A

species reactivity

human, mouse, rat

species reactivity (predicted by homology)

monkey

packaging

antibody small pack of 25

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

affinity binding assay: suitable, immunohistochemistry: suitable, western blot: suitable

isotype

IgG

epitope sequence

Internal

Protein ID accession no.

UniProt accession no.

shipped in

ambient

storage temp.

2-8°C

Quality Level

Gene Information

human ... OLR1(4973)

Preparation Note

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.
300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Application

Quality Control Testing

Evaluated by Western Blotting in RAW 264.7 cell lysate.

Western Blotting Analysis: 1:1,000 dilution of this antibody detected OLR1 in RAW 264.7 cell lysate.

Tested Applications

Western Blotting Analysis: 1:1,000 dilution from a representative lot detected OLR1 in human, mouse, and rat kidney tissue lysates.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected OLR1 in human, rat, and mouse kidney tissue sections.

Affinity Binding Assay: A representative lot of this antibody bound with OLR1 peptide with at least one thousand-fold (1,000X) higher affinity than with non-specific control peptide.

Note: Actual optimal working dilutions must be determined by end user as specimens and experimental conditions may vary with the end user.

Biochem/physiol Actions

Clone 2E2 is a ZooMAb® rabbit recombinant monoclonal antibody that detects OLR1. It targets an epitope within 16 amino acids from the extracellular domain.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

General description

ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Oxidized low-density lipoprotein receptor 1 (UniProt: P78380; also known as Ox-LDL receptor 1, C-type lectin domain family 8 member A, Lectin-like oxidized LDL receptor 1 (LOX-1; Lectin-like oxLDL receptor 1; hLOX-1), Lectin-type oxidized LDL receptor 1, Oxidized low-density lipoprotein receptor 1, soluble form) is encoded by the OLR1 (also known as CLEC8A, LOX1) gene (Gene ID: 4973) in human. Oxidized low-density lipoprotein receptor 1 (OLR1), also known as LOX-1, is a transmembrane protein belonging to the C-type lectin superfamily. Structurally, it consists of a short intracellular domain, a transmembrane region, and an extracellular lectin-like domain that facilitates binding to modified lipoproteins, such as oxidized LDL (oxLDL). This receptor plays a critical role in vascular biology by mediating the recognition, internalization, and degradation of oxLDL, which is central to its function as a scavenger receptor. OLR1 is localized to the plasma membrane, nucleoplasm, and vesicles, indicating its dynamic role in cellular processes. OLR1 is involved in inflammation, oxidative stress, and apoptosis. It regulates pathways such as nuclear factor- B (NF- B) signaling, which drives the expression of pro-inflammatory cytokines and adhesion molecules. OLR1 expression is typically low under physiological conditions but is upregulated in response to atherosclerotic stimuli, including tumor necrosis factor-alpha (TNF- ), shear stress, and oxLDL exposure. This upregulation enhances endothelial dysfunction and promotes atherogenesis. OLR1 also plays roles in other pathological contexts, including ischemic stroke, diabetes, and cancer, where its activity contributes to oxidative stress and inflammation. OLR1 interacts with various proteins and signaling pathways, including those involved in lipid metabolism, immune cell activation, and apoptosis. Its expression is regulated by cyclic AMP signaling and is influenced by inflammatory mediators. Mutations in the OLR1 gene have been associated with increased susceptibility to myocardial infarction and other cardiovascular diseases. Soluble OLR1 (sOLR1), which is derived from proteolytic cleavage of the membrane-bound receptor, serves as a biomarker for vascular inflammation and atherosclerosis. This ZooMAb® recombinant monoclonal antibody, generated by our proprietary technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.

Immunogen

KLH-conjugated linear peptide corresponding to 16 amino acids from the extracellular domain of human OLR1.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service