MilliporeSigma
All Photos(1)

11325-U

Supelco

Carbotrap® Adsorbent

matrix Carbotrap® B, 20-40 mesh, pack of 144 × 190 mg

EC Number:
NACRES:
SB.54

Quality Level

product line

Carbotrap®

form

granular

packaging

pack of 144 × 190 mg

technique(s)

LPLC: suitable

matrix

Carbotrap® B

matrix active group

carbon

particle size

20-40 mesh

pore size

~0 cm3/g macroporosity
~0 cm3/g mesoporosity
~0 cm3/g microporosity
~0 Å pore diameter

surface area

~100 m2/g

density

~0.38 g/mL (free fall density)

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

Carboxen® 569 was used for trapping a range of replacement chlorofluorocarbons varying in boiling point from -48.4 to -9.8°C which is highly advantageous for analysis of trace gases in atmosphere.
Carboxen® Adsorbent is ideally used for solid-phase microextraction (SPME). It is highly efficient in extraction of small polar molecules from aqueous solution.
A graphitized carbon black (GCB) can be non-porous or porous. The graphitization process results in a highly pure surface with great adsorption and desorption (if required) properties. Surface interactions depend solely on dispersion (London) forces. These particles are:
  • Granular
  • Friable
  • Used for molecules with an analyte size relative to C3-C20+ n-alkanes
  • Hydrophobic (can be used in high humidity environments)

Generally, GCB adsorbents offer weaker relative adsorptive strength compared to carbon molecular sieve (CMS) adsorbents, and similar relative adsorptive strength compared to spherical graphitized polymer carbon (SGPC) adsorbents. Our Carbotrap products are a type of GCB adsorbent.
  • Particles are 20/40 mesh
  • These large particles allow high flow rates without excessive pressure drops

For more information about any of our specialty carbon adsorbents, please visit sigma-aldrich.com/carbon

Footnote

Pack of 144 ampules

Legal Information

Carbotrap is a registered trademark of Sigma-Aldrich Co. LLC
Carboxen is a registered trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Articles

High Purity Carbon Adsorbents for Sample Preparation and Chromatographic Applications

Carbon Molecular sieves (CMS) are a versatile range of adsorbents that can be tailored for specific applications. Supelco® scientists have been synthesizing synthetic CMS carbons for several decades, starting from tailoring of the starting polymers/copolymers, to modifying the final properties of the subsequent CMS carbon.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service