19134-U

pk6 Needles 26s ga 2in Pt2 250uL-10mL

needle size 26s ga, needle L 51 mm (2 in.), bevel tip stainless steel needle (point style 2), pkg of 6 ea

Synonym(s):
Hamilton needles for RN syringes
NACRES:
NB.03

material

bevel tip stainless steel needle (point style 2)

needle size

26s ga

packaging

pkg of 6 ea

manufacturer/tradename

Hamilton 7779-02

needle L

51 mm (2 in.)

volume

250-10000 μL

compatibility

for use with 250μL-10mL syringe

Looking for similar products? Visit Product Comparison Guide

Related Categories

Legal Information

Hamilton is a registered trademark of Hamilton Co.

RIDADR

NONH for all modes of transport

WGK Germany

3

Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.