All Photos(1)

20077-U

Supelco

Modified BTEXTRAP M, Tekmar® 2000

Synonym(s):
Modified BTEX Trap M
NACRES:
NB.21

application(s)

environmental

Quality Level

compatibility

for use with Tekmar® Velocity XPT, LSC-1, LSC-2, LSC-2000, 4000 (Straight, 12 in./30.5 cm, Swagelok fitting), HT3

Looking for similar products? Visit Product Comparison Guide

Related Categories

Legal Information

BTEXTRAP is a trademark of Sigma-Aldrich Co. LLC
Tekmar is a registered trademark of Tekmar Co.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service