Documents
20845
Supelco
Empty Glass GC Column, for Varian®, Universal for 3000, 4000, 6000, and Vista (On-Column)
L × O.D. × I.D. 6.0 ft (1.8 m) × 1/4  in. × 2.0 mm
Pricing and availability is not currently available.
material
glass column
application(s)
gas chromatography (GC): suitable
L × O.D. × I.D.
6.0 ft (1.8 m) × 1/4  in. × 2.0 mm
column type
empty
General description
GC columns are of two types; packed column and capillary column. Capillary column carries the gas through a central aperture which is unimpeded through the entire length of column. In general, capillary columns are made longer to produce more theoretical plates. This also helps in band dispersion eventually narrowing the peaks and resulting in high resolution.
Silane treated empty glass column. Fittings are not included.
  • X (length of injector arm) = 9.25 in. (235 mm)
  • Y (length of detector arm) = 2.0 in. (51 mm)
  • S (span, injector to detector) = n/a
This column fits Varian 3000, 4000, 6000, and Vista (on-column injection). The three piece construction (injector arm + coil, union, and detector arm) allows its use in any injector to detector position (A to A, B to B, A to B, and B to A).
Application
Empty column may be used for packing adsorbents for temperature-programmed desorption (TPD) instrumentation.
Legal Information
Varian is a registered trademark of Varian Medical Systems, Inc.
Vista is a trademark of Varian Medical Systems, Inc.
Certificate of Analysis
Certificate of Origin
The determination of active filter aid adsorption sites by temperature-programmed desorption.
Zhu. Z. Y.
Journal of the American Oil Chemists' Society, 71, 189-194 (1994)
Raymond P. W. Scott
Introduction to Analytical Gas Chromatography (1997)
Contact Technical Service
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.