MilliporeSigma
All Photos(2)

20875

Supelco

Carbotrap® 300

glass TD tube, fritted, O.D. × I.D. × L 6 mm × 4 mm × 4.5 in., preconditioned, pkg of 1 ea

material

glass TD tube

Agency

EPA TO-1,TO-2,TO-3,TO-17

description

Sealed with TDS³ Storage Container

feature

fritted
preconditioned

packaging

pkg of 1 ea

O.D. × I.D. × L

6 mm × 4 mm × 4.5 in.

matrix

Carbosieve S-III carbon molecular sieve (CMS)
Carbotrap® 300 (Packed with Carbotrap C, Carbotrap B, Carbosieve SIII)
Carbotrap® B graphitized carbon black (GCB)
Carbotrap® C graphitized carbon black (GCB)

application(s)

environmental
industrial hygiene

compatibility

for use with Dynatherm ACEM-9300, ACEM-900 850/890 TDU, MTDU, and OI Analytical Eclipse Air Tube Desorber

Looking for similar products? Visit Product Comparison Guide

Application

C2 and larger compounds in air

Legal Information

Carbosieve is a trademark of Sigma-Aldrich Co. LLC
Carbotrap is a registered trademark of Sigma-Aldrich Co. LLC
Dynatherm is a trademark of CDS Analytical

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service