Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
41121800
NACRES:
SB.54
material
glass column
technique(s)
gas chromatography (GC): suitable
L × O.D. × I.D.
6.0 ft (1.8 m) × 1/4 in. × 4.0 mm
matrix active group
Empty Glass Column
column type
empty
compatibility
for use with Varian
General description
Silane treated empty glass column. Fittings are not included.
- X (length of injector arm) = 9.31 in. (236 mm)
- Y (length of detector arm) = 8.0 in. (203 mm)
- S (span, injector to detector) = 5.5 in. (140 mm)
GC columns are of two types; packed column and capillary column. Capillary column carries the gas through a central aperture which is unimpeded through the entire length of column. In general, capillary columns are made longer to produce more theoretical plates. This also helps in band dispersion eventually narrowing the peaks and resulting in high resolution.
Application
Empty column may be used for packing adsorbents for temperature-programmed desorption (TPD) instrumentation.
Legal Information
Varian is a registered trademark of Varian Medical Systems, Inc.
Still not finding the right product?
Explore all of our products under Empty Glass GC Column, for Varian® 3000, 4000, 6000, and Vista (FID, Inj. A to Det. A, On-Column)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.