22136-U

Swagelok® Plug

Swagelok®, 100-P, brass, 1/16 in. Swagelok

Synonym(s):
Swagelok 100P,BRASS PLUGS 1/16 PK3
NACRES:
SB.54
Pricing and availability is not currently available.

material

brass

packaging

pkg of 3 ea

mfr. no.

Swagelok®, 100-P

fitting

1/16 in. Swagelok

Looking for similar products? Visit Product Comparison Guide

Application

Brass Swagelok® plugs maybe used to close the sample port during chromatographic analysis of programmed-temperature pyrolysis products.

Legal Information

Swagelok is a registered trademark of Swagelok Company

RIDADR

NONH for all modes of transport

WGK Germany

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis
Certificate of Origin
Combined pyrolysis and thin-layer chromatography. Study of decomposition mechanisms.
Raymon RN.
Analytical Chemistry, 39 (7), 730-733 (1967)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.