Documents

22700-U

Swagelok® Connector to Male NPT

stainless steel, 1/4 in. Swagelok, 1/4 in. male NPT, 400-1-4

Synonym(s):
Swagelok 400-1-4,SS CONNECTOR 1/4TX1/4P EA
eCl@ss:
32035302
NACRES:
SB.54
Pricing and availability is not currently available.

material

stainless steel

packaging

pkg of 1 ea

mfr. no.

, 400-1-4

fitting

1/4 in. Swagelok
1/4 in. male NPT

Looking for similar products? Visit Product Comparison Guide

General description

Nuts and ferrules included.

Legal Information

Swagelok is a registered trademark of Swagelok Company

RIDADR

NONH for all modes of transport

WGK Germany

3

Certificate of Analysis
Certificate of Origin
Gas Chromatographic Analysis of C1 to C4 Hydrocarbons in the Parts per Million Range in Air and in Vaporized Liquid Oxygen.
Kuley, C. J.
Analytical Chemistry, 35, 1472-1475 (1963)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.