24764
Supelco
Screw cap (open-top), polypropylene
black polypropylene cap, 8-425 thread for cap size, pkg of 1000 ea
Pricing and availability is not currently available.
material
black polypropylene cap
feature
autoclavable
closure type black polypropylene hole cap
temp. limit 135 °C
packaging
pkg of 1000 ea
fitting
8-425 thread for cap size
compatibility
for use with Screw Top, 2 mL, 4.6 mm I.D.
Related Categories
General description
These hole caps do not come with septa.
Certificate of Analysis
Certificate of Origin
Contact Technical Service
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.