26724
Syringe for On-column Injection, Removable needle
701RN, volume 10 μL, needle L × O.D. 12.5 cm × 0.23 mm
eCl@ss:
32070500
Pricing and availability is not currently available.
material
bevel tip stainless steel needle
glass syringe
packaging
pkg of 1 ea
mfr. no.
Hamilton, 80386
needle L × O.D.
12.5 cm × 0.23 mm
volume
10 μL
Related Categories
General description
Use the 701RNFS syringes with liquid samples. A PTFE plunger tip makes the 1701RNFS syringes gas tight. Syringes come with a replaceable fused silica needle. Capacity 10 μL, 0.17 mm OD, 10 cm length, point style #3. Use with 0.20 mm I.D. or larger columns.
Certificate of Analysis
Certificate of Origin
Contact Technical Service
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.