27084-U

Supelco

Vials, screw top, amber glass (vial only)

volume 2 mL, amber glass vial (with marking spot), thread for 8-425, pkg of 100 ea

NACRES:
NB.11
Pricing and availability is not currently available.

material

amber glass vial (with marking spot)

feature

closure type screw top vial
marking spot

packaging

pkg of 100 ea

O.D. × H × I.D.

12 mm × 32 mm × 4.6 mm

volume

2 mL

fitting

thread for 8-425

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Amber glass vials (with marking spot, volume 2 mL) are useful in preventing the sensitive analyte under investigation from exposure to UV light. Screw top vials are used on many autosamplers with a standard opening of 4.6 mm. The threads on the standard vials are 8-425.

Legal Information

WHEATON is a registered trademark of Wheaton Industries, Inc.

WGK Germany

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis
Certificate of Origin
Eman Ibrahim et al.
Biotechnology for biofuels, 10, 52-52 (2017-04-18)
The high crystallinity of cellulosic biomass myofibrils as well as the complexity of their intermolecular structure is a significant impediment for biofuel production. Cloning of β-Jelly roll topology, (α/α) This study provides some insight into which parameters should be optimized...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.