Documents

27087-U

Supelco

Vials, screw top, amber glass (vial only)

volume 2 mL, amber glass vial, thread for 8-425, pkg of 1000 ea

NACRES:
NB.11
Pricing and availability is not currently available.

material

amber glass vial

feature

closure type screw top vial

packaging

pkg of 1000 ea

O.D. × H × I.D.

12 mm ≤ × 32 mm × 4.6 mm

volume

2 mL

fitting

thread for 8-425

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Vials are suitable basically to inject samples from an autosampler. They are available in various sizes, although 2 mL vials are more commonly used. Autosampler needle pierces through the cap during injection and withdraws the required aliquots of sample from the vials. Amber glass vials (volume 2 mL) are useful in preventing the sensitive analyte under investigation from exposure to UV light. Screw top vials are used on many autosamplers with a standard opening of 4.6 mm. The threads on the standard vials are 8-425.

WGK Germany

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.