27478-U

Supelco

Micro Reaction vessel, amber glass, pkg of 12

volume 5 mL, O.D. × H 20 mm × 62 mm, thread for 20-400

NACRES:
NB.11

material

amber glass vial

feature

closure type black phenolic hole cap

packaging

pkg of 12 ea

O.D. × H

20 mm × 62 mm

volume

5 mL

fitting

thread for 20-400

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Cap and septa for the amber Micro Reaction Vessels are as follows:
  • 1mL (13/425 thead): Hole Cap 27120-U, Septa 27145
  • 2-5 mL (20-400 thread): Hole Cap 27176, Septa 27178

RIDADR

NONH for all modes of transport

WGK Germany

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.