MilliporeSigma
  • 303100U
All Photos(1)

303100-U

Supelco

GPA Natural Gas Reference Standard

pkg of 785 g, volume 14 L, analytical standard

Synonym(s):
Natural Gas Reference Standard
NACRES:
NA.24

grade

analytical standard

Quality Level

description

GPA

form

gas

packaging

pkg of 785 g

manufacturer/tradename

Scott Gas TB000010-14

availability

not available in EU

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

volume

14 L

format

multi-component solution

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

1 of 4

This Item
29564-U29556-U29575-U
grade

analytical standard

grade

-

grade

-

grade

-

form

gas

form

-

form

-

form

-

packaging

pkg of 785 g

packaging

-

packaging

-

packaging

-

manufacturer/tradename

Scott Gas TB000010-14

manufacturer/tradename

-

manufacturer/tradename

-

manufacturer/tradename

-

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

-

technique(s)

-

technique(s)

-

format

multi-component solution

format

-

format

-

format

-

Application

GPA Natural Gas Reference Standard is a gas calibration standard, suitable for GC calibration, testing of fixed and personal gas monitors, and testing of in situ industrial stack analyzers. A certificate of analysis is included.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Features and Benefits

  • GPA Natural Gas Reference Standard is packaged in lightweight, non-refillable, recyclable containers.
  • Portable, convenient, and easy to use.
  • Manufactured gravimetrically for maximum accuracy.
  • NIST traceable.

Other Notes

This cylinder is not "TC" compliant and cannot be sold in Canada.

Analyte

Description

    Helium .5 mol %
    Nitrogen 5 mol %
    Carbon dioxide 1 mol %
    Methane 70.5 mol %
    Ethane 9 mol %
    Propane 6 mol %
    2-Methylpropane 3 mol %
    Butane 3 mol %
    2-Methylbutane 1 mol %
    Pentane 1 mol %
    See All (10)

Pictograms

FlameGas cylinder

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Gas 1A - Press. Gas Compr. Gas

Storage Class Code

2A - Gases

WGK

WGK 2

Flash Point(F)

-306.4 °F - closed cup

Flash Point(C)

-188 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service