MilliporeSigma
All Photos(1)

442533

Supelco

Cyclohexanone-2,4-DNPH

analytical standard

Synonym(s):
Cyclohexanone 2,4-dinitrophenylhydrazone
Linear Formula:
C6H10[=NNHC6H3(NO2)2]
CAS Number:
Molecular Weight:
278.26
EC Number:
MDL number:
PubChem Substance ID:

grade

analytical standard

feature

standard type calibration

packaging

ampule of 100 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

159-160 °C (lit.)

application(s)

environmental

format

neat

storage temp.

room temp

SMILES string

[O-][N+](=O)c1ccc(N\N=C2\CCCCC2)c(c1)[N+]([O-])=O

InChI

1S/C12H14N4O4/c17-15(18)10-6-7-11(12(8-10)16(19)20)14-13-9-4-2-1-3-5-9/h6-8,14H,1-5H2

InChI key

QLWXZRVOHCYKKK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service